missing translation for 'onlineSavingsMsg'
Learn More
3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, 99%, Thermo Scientific Chemicals Product Code.: 10229550

3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, 99%, Thermo Scientific Chemicals

Product Code. 10229550
500 mg, Glass bottle
Click to view available options
Quantity:
500 mg
2 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10229550

Brand: Thermo Scientific Acros 401105000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34272-83-8
Molecular Formula C8H17N2O
Molecular Weight (g/mol) 157.24
MDL Number MFCD00046088
InChI Key USGBLEMQARVRDC-UHFFFAOYSA-N
Synonym 3s-1-hydroxy-2,2,5,5-tetramethylpyrrolidin-3-aminium
PubChem CID 558590
IUPAC Name 1-hydroxy-2,2,5,5-tetramethylpyrrolidin-3-amine
SMILES CC1(CC(C(N1O)(C)C)N)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 45.0°C
Color Orange
Boiling Point 75.0°C to 80.0°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 500 mg
Formula Weight 157.24
Percent Purity 99%
Physical Form Hygroscopic Crystals
Chemical Name or Material 3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459030 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number401105000,401100020
Lot NumberA0459030 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number401105000,401100020
Lot NumberA0403310 Certificate TypeCertificate of Analysis Date05/12/2022 Catalog Number401105000,401100020
compliance-icons compliance-icons
Product Identifier
  • 3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.