missing translation for 'onlineSavingsMsg'
Learn More
3-Isochromanone, Thermo Scientific Chemicals Product Code.: 17221146

3-Isochromanone, Thermo Scientific Chemicals

Product Code. 17221146
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 17221146

Brand: Thermo Scientific Acros 370130010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4385-35-7
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
InChI Key ILHLUZUMRJQEAH-UHFFFAOYSA-N
IUPAC Name 3,4-dihydro-1H-2-benzopyran-3-one
SMILES O=C1CC2=CC=CC=C2CO1

Specifications

Melting Point 83.0°C to 86.0°C
Quantity 1 g
Beilstein 17, V,10, 19
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in methanol
Formula Weight 148.16
Chemical Name or Material 3-Isochromanone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0424383 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number370130050,370130250,370130010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Isochromanone, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.