Learn More
Euro Protection™ 3D Flat Fold Masks FFP3 NR D
Euro Protection™ 3D Flat Fold Masks FFP3 NR D are a single-use protective mask with a nasal clip and elastic for a secure, and comfortable fit.
Brand:  Euro Protection™ 23385
Product Code. 15975920
  • £68.28 / Pack of 20
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Multi-layer design
  • Soft inner layer
  • Adjustable tightening elastic
  • Nose clip
  • Nose seal/face seal
Specifications
Specifications
EN 149:2001 + A1:2009
SMS Polypropylene
Melt-Blown electrostatic polypropylene
Yes
Soft polypropylene
Nose clip
Yes
Single-use
One Size
Nose clip
Latex-free synthetic rubber
Reinforced Polyethylene
20/Box, 12 Boxes/Cs.
18 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Euro Protection™ 3D Flat Fold Masks FFP3 NR D > 20/Box, 12 Boxes/Ctn