Learn More
4-Nitrophenylphosphorylcholine, Thermo Scientific Chemicals
Description
O-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of Phospholipase C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
O-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of Phospholipase C.
Solubility
Soluble in water
Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Melting Point | 233°C to 235°C |
Color | Yellow |
Quantity | 100 mg |
Beilstein | 1895422 |
Sensitivity | Light and moisture sensitive |
Solubility Information | Soluble in water |
Formula Weight | 304.24 |
Physical Form | Solid |
Chemical Name or Material | 4-Nitrophenylphosphorylcholine |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N19L004 | Certificate of Analysis | 21/02/2025 | J66465.ME,J66465.03,J66465.MC |
W11K013 | Certificate of Analysis | 23/09/2024 | J66465.ME,J66465.03,J66465.MC |
N07K023 | Certificate of Analysis | 14/02/2024 | J66465.ME,J66465.03,J66465.MC |
Z09J009 | Certificate of Analysis | 28/12/2023 | J66465.ME,J66465.03,J66465.MC |
Y09J019 | Certificate of Analysis | 05/12/2023 | J66465.ME,J66465.03,J66465.MC |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.