missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Lecithin, Refined Product Code.: 11367948

Thermo Scientific Chemicals Lecithin, Refined

Product Code. 11367948
250 g
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11367948

Brand: Thermo Scientific Chemicals 036486.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lecithin is used as an emulsifying and stabilizing agent in the food, pharmaceutical, and cosmetic industries. Lecithin is taken as a medicine and is also used in the manufacturing of medicines. Soybean lecithin is useful for its application in liquid ram semen. Fractionation process of sunflower lecithin with absolute ethanol was used for obtaining enriched fractions in certain phospholipids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8002-43-5
Molecular Formula C42H80NO8P
Molecular Weight (g/mol) 758.075
MDL Number MFCD00147406
InChI Key JLPULHDHAOZNQI-JLOPVYAASA-N
Synonym 3-sn-phosphatidylcholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholineShow MoreShow Less
PubChem CID 16213884
ChEBI CHEBI:86658
IUPAC Name [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC

Specifications

Quantity 250 g
Beilstein 5209585
Solubility Information Soluble in chloroform. Very partially soluble in water.
Physical Form Powder
Chemical Name or Material Lecithin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN21L034 Certificate TypeCertificate of Analysis Date24/02/2025 Catalog Number036486.A1,036486.30
Lot NumberZ18K012 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog Number036486.A1,036486.30
Lot NumberW17K058 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog Number036486.A1,036486.30
Lot NumberU20K038 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number036486.A1,036486.30
Lot NumberS19K028 Certificate TypeCertificate of Analysis Date25/06/2024 Catalog Number036486.A1,036486.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Lecithin, Refined >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.