missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 43.2mm Septum for SCHOTT Screw Caps
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
Brand: Fisherbrand™ 43 02 1600
Description
- For use with SCHOTT Screw Caps

Specifications
Septa | |
43.2 mm | |
500 |
Gray, Red | |
Butyl, PTFE | |
1.6 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ 43.2mm Septum for SCHOTT Screw Caps > Butyl red/PTFE grey,1.6 mm thickness,55° shore A
Spot an opportunity for improvement?Share a Content Correction