missing translation for 'onlineSavingsMsg'
Learn More
5-Methyl-1H-benzotriazole, 98+%, Thermo Scientific Chemicals Product Code.: 11443314

5-Methyl-1H-benzotriazole, 98+%, Thermo Scientific Chemicals

Product Code. 11443314
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11443314

Brand: Thermo Scientific Alfa Aesar A10615.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.

Solubility
Soluble in chloroform, methanol, water (6.0 g/L at 25°C), ethanol, and toluene (slightly).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-85-6
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005702
InChI Key LRUDIIUSNGCQKF-UHFFFAOYSA-N
Synonym 5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1Show MoreShow Less
PubChem CID 8705
ChEBI CHEBI:83455
IUPAC Name 5-methyl-2H-benzotriazole
SMILES CC1=CC2=NNN=C2C=C1

Specifications

Melting Point 77°C to 84°C
pH 5.3
Boiling Point 210°C to 212°C (12 mmHg)
Odor Odorless
Quantity 25 g
Beilstein 116658
Solubility Information Soluble in chloroform,methanol,water (6.0g/L at 25°C),ethanol,and toluene (slightly).
Formula Weight 133.15
Percent Purity ≥98%
Chemical Name or Material 5-Methyl-1H-benzotriazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS13K054 Certificate TypeCertificate of Analysis Date03/07/2024 Catalog NumberA10615.22,A10615.14,A10615.36
Lot NumberN09K017 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog NumberA10615.22,A10615.14,A10615.36
Lot NumberN14J050 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA10615.22,A10615.14,A10615.36
Lot NumberR12I015 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog NumberA10615.22,A10615.14,A10615.36
Lot NumberT18I008 Certificate TypeCertificate of Analysis Date27/07/2022 Catalog NumberA10615.22,A10615.14,A10615.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Methyl-1H-benzotriazole, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.