missing translation for 'onlineSavingsMsg'
Learn More
6-Chloro-1-benzothiopyran-4-one 1,1-dioxide, 97%, Maybridge™ Product Code.: 10425332

6-Chloro-1-benzothiopyran-4-one 1,1-dioxide, 97%, Maybridge™

Product Code. 10425332
1 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 10425332

Brand: Thermo Scientific Maybridge SEW00597DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 90396-06-8
Molecular Formula C9H7ClO3S
Molecular Weight (g/mol) 230.66
MDL Number MFCD00052999
InChI Key ZFZPMYLZPMMUQA-UHFFFAOYSA-N
Synonym 6-chloro-1-benzothiopyran-4-one 1,1-dioxide, 6-chloro-2,3-dihydro-1??-benzothiopyran-1,1,4-trione, maybridge1_007519, 6-chlorothiochroman-4-one 1,1-dioxide, 6-chloro-2h,3h-benzo e thiin-1,1,4-trione, 6-chloro-1,1-dioxo-1lambda6-thiochroman-4-one, 6-chloro-2,3-dihydro-1-benzothiopyran-1,1,4-trione, 6-chloro-2,3-dihydro-4h-thiochromen-4-one 1,1-dioxide, 6-chloro-2,3-dihydro-1, e6-benzothiopyran-1,1,4-trione, 6-chloro-3,4-dihydro-2h-1??-benzothiopyran-1,1,4-trioneShow MoreShow Less
PubChem CID 2739106
IUPAC Name 6-chloro-1,1-dioxo-2,3-dihydrothiochromen-4-one
SMILES ClC1=CC=C2C(=C1)C(=O)CCS2(=O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 230.67
Percent Purity 97%
Chemical Name or Material 6-Chloro-1-benzothiopyran-4-one 1, 1-dioxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Chloro-1-benzothiopyran-4-one 1,1-dioxide, 97%, Maybridge™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.