Learn More
Thermo Scientific™ 6-Place Fixed-Angle Highconic™ Rotor Adapters
Use these rotor adapters to connect with round or conical-bottom centrifuge tubes and microtubes tubes up to 50mL.
Brand:  Thermo Scientific™ 75003102
 View more versions of this product
Product Code. 10676373
  • £79.23 / Each
Explore more special offers
Requires : 50mL conical adapter 75-007-556 requires rubber pads (supplied with buckets). Remove pads when using all other size adapters.
Description
Description
Use these 6-Place Fixed-Angle Highconic™ Rotor Adapters to connect with centrifuge tubes up to 50mL. The adapters accommodate round or conical-bottom tubes and microtubes and work with HiGhConic™ fixed angle rotors.
  • The adapters accommodate round- or conical-bottom tubes and microtubes.
  • Rotor adaptors work with HiGhConic™ fixed angle rotors
  • Fits tube sizes up to 50mL
Order Info
Adapters are sold individually; order correct quantity to equip rotor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotor Adapter
50 mL
50 mL
Conical
HiGhConic™ fixed angle rotor
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ 6-Place Fixed-Angle Highconic™ Rotor Adapters > 50mL