missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 9mm Short Thread Glass Vial, Wide Opening, Amber
13.16 GBP valid until 2025-12-31
Use promo code "24355" to get your promotional price.
Description
- 9mm
- Short Thread
- Glass
- Wide opening
- Amber

Specifications
SureStop | |
11.6 mm | |
1.85 mL | |
Short Thread | |
<120 μL | |
Flat Bottom | |
100 | |
No |
Amber | |
Glass | |
GC, HPLC | |
CleanPack, Polypropylene Box | |
1.5 mL | |
32 mm | |
ND9 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 15
Special Offers
-
Multiple Manufacturers
Incredible Savings on Peptide and Protein Mass Spectrometry Testing Products
Explore our new Peptide and Protein Mass Spectrometry Testing Products PromotionPromo Code: 24142Expires: 2025-03-14
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
47Hours
0Minutes
36Seconds
38 -
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
17Hours
0Minutes
36Seconds
38
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ 9mm Short Thread Glass Vial, Wide Opening, Amber > SureStop,flat bottom,1.5ml
Spot an opportunity for improvement?Share a Content Correction