Learn More
Thermo Scientific™ Agarase (0.5 U/μL)
Specifically digests the agarose polysaccharide core, which allows for gentle yet efficient recovery of DNA or RNA fragments from low melting point agarose.
Brand: Thermo Scientific™ EO0461
89.06 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Agarase specifically digests the agarose polysaccharide core into neoagaro-oligosaccharides. Agarase allows for gentle yet efficient recovery of DNA or RNA fragments from low melting point agarose. The recovered nucleic acids can be directly used for amplification, cloning, sequencing, etc.
Highlights
- Gentle -recovers intact DNA and RNA
- High recovery yields -> 90% of the starting DNA recovered following extraction
- Ideal for both long (> 30 kb) and short (< 100 bp) DNA fragments
- Active in TAE and TBE electrophoresis buffers

Specifications
0.5 U/μL | |
Electrophoresis Buffer | |
Agarose |
Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), PCR, Cloning, cDNA Library Construction, Sequencing, In Vitro Transcription, Nuclease Protection Assays, Nucleic Acid Labeling | |
100 units |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
86Hours
5Minutes
31Seconds
10New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.