missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Alkaline Phosphatase Buffer-1 (5X)

For cellular and molecular biology applications

Brand:  Thermo Scientific™ J62907.AK

60.70 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15415209

  • £61.00 / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Formulation: 200mM Tris, 200mM NaCl, and 40mM magnesium chloride at pH 9.5
TRUSTED_SUSTAINABILITY
Specifications

Specifications

200mM Tris, 200mM NaCl, and 40mM magnesium chloride at pH 9.5
250 mL
Liquid
Keep cold
Alkaline Phosphatase Buffer-1 (5X)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Alkaline Phosphatase Buffer-1 (5X) > 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only