missing translation for 'onlineSavingsMsg'
Learn More
Ansell™ AlphaTec™ 38-612 (ex ChemTek™ 38-612) Product Code.: 15886932
Add to basket

Ansell™ AlphaTec™ 38-612 (ex ChemTek™ 38-612)

Product Code. 15886932 Shop All Ansell Products
8
Click to view available options
Size:
8
9

Product Code. 15886932

Brand: Ansell™ 38612080

Add to basket
Add to basket

Maximum protection from hazardous chemicals

  • The AlphaTec™ 38-612 range provides the best resistance to the most aggressive chemicals – without compromising dexterity or comfort.
  • AlphaTec™ 38-612 offers protection against aliphatic, halogenated and aromatic hydrocarbons as well as concentrated mineral acids.
  • AlphaTec™ 38-612 gloves very flexible and provide excellent dexterity. Designed for comfort – with a natural, curved ergonomic shape and soft feel – they offer easy donning and good grip.
TRUSTED_SUSTAINABILITY

Specifications

Cut Style Beaded
Coating Material Viton/Butyl Polymer
Color Black
Exterior Finish Smooth
Latex Free Yes
Material Butyl Polymer
Size 8
Thickness (Metric) 0.3 mm
Length (English) 12 in.
Length (Metric) 300 mm
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131

    Expires2025-06-08

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell™ AlphaTec™ 38-612 (ex ChemTek™ 38-612) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.