Learn More
Ansell™ AlphaTec™MICROCHEM™4000 Model 215 Apron with Sleeves Product Code.: 15895342
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
Ansell™ AlphaTec™MICROCHEM™4000 Model 215 Apron with Sleeves
Product Code. 15895342 Shop All Ansell Products
3X-Large
Click to view available options
Size:
Small
Medium
Large
2X-Large
3X-Large
4X-Large
Product Code. 15895342
Brand: Ansell™ GR40T0021506
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
Add to basket
MICROCHEM™ 4000 is designed to provide anexceptional barrier against organic and inorganicchemicals and biological agents
  • Velcro fastening to neck
  • Tie fastening at the waist
  • Double cuff with knitted
  • inner cuff
  • Welded and taped seams
TRUSTED_SUSTAINABILITY
Specifications
Size 3X-Large
Material Multi-layer Barrier Laminate Fabric
Color Green
Closure Type Hook and Loop, Waist Ties
Sterility Non-sterile
Cuff Style Double Inner Knit
Latex Free Yes
No. of Pockets None
Resistance Abrasions, acids, anti-static, bloodborne pathogens, chemicals
Certifications/Compliance EN 14605: 2005 +A1:2009, Type PB (3); EN 14605: 2005 +A1:2009, Type PB (4); EN14126: 2003, Types PB(3)-B, PB (4)-B
Style Alphatec™ 4000, Model 215
Seam Type Welded and Taped
Show More Show Less
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
Ansell™ AlphaTec™MICROCHEM™4000 Model 215 Apron with Sleeves >