missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Xtra SIL G Silica Layers on Alugram™ Aluminum Sheets UV254

Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Xtra SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.

£24.13 - £254.00

Specifications

Particle Size 5 to 17 μm
Pore Size 60 Å
Type Aluminum Sheet
Fluorescent Indicator UV254
Pore Volume 0.75 mL/g
View More Specs

Products 6
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
10707898
View Documents
Macherey-Nagel™
818362
10 x 20 cm
£86.75
Pack of 20
In Stock
to see stock.
Add to basket
 
12804281
View Documents
Macherey-Nagel™
818333
20 x 20 cm
£254.00
Pack of 25
In Stock
to see stock.
Add to basket
 
12760763
View Documents
Macherey-Nagel™
818331
4 x 8 cm
£69.25
Pack of 50
In Stock
to see stock.
Add to basket
 
12780813
View Documents
Macherey-Nagel™
818360
5 x 10 cm
£104.00
Pack of 50
In Stock
to see stock.
Add to basket
 
12730773
View Documents
Macherey-Nagel™
818332
5 x 20 cm
£139.00
Pack of 50
In Stock
to see stock.
Add to basket
 
11239889
View Documents
Macherey-Nagel™
818330.20
5 x 7.5 cm
£24.13
Pack of 20
In Stock
to see stock.
Add to basket
 
Description

Description



  • Unmodified standard silica layers on aluminum for TLC
  • Silica 60
  • Outstanding wettability for precise colorization results, even with 100 % aqueous detection reagents
  • Excellent separation efficiency and reproducibility from lot to lot
  • Easy and reliable cutting due to an optimized binder system, no flaking of silica
  • Binder: highly polymeric product, which is stable in almost all organic solvents and resistant towards aggressive visualization reagents; also completely stable in purely aqueous eluents
Specifications

Specifications

5 to 17 μm
Aluminum Sheet
0.75 mL/g
0.20 mm
60 Å
UV254
∼500 m2/g
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ Xtra SIL G Silica Layers on Alugram™ Aluminum Sheets UV254

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.