Learn More
Macherey-Nagel™ Xtra SIL G Silica Layers on Alugram™ Aluminum Sheets UV254
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Xtra SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.
Brand: Macherey-Nagel™ 818362
81.15 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Unmodified standard silica layers on aluminum for TLC
- Silica 60
- Outstanding wettability for precise colorization results, even with 100 % aqueous detection reagents
- Excellent separation efficiency and reproducibility from lot to lot
- Easy and reliable cutting due to an optimized binder system, no flaking of silica
- Binder: highly polymeric product, which is stable in almost all organic solvents and resistant towards aggressive visualization reagents; also completely stable in purely aqueous eluents
Specifications
5 to 17 μm | |
20 Pk. | |
UV254 | |
∼500 m2/g | |
10 x 20 cm |
60 Å | |
Aluminum Sheet | |
0.75 mL/g | |
0.20 mm |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.