missing translation for 'onlineSavingsMsg'
Learn More
Aluminum oxalate hydrate, 99%, Thermo Scientific Chemicals Product Code.: 11369648

Aluminum oxalate hydrate, 99%, Thermo Scientific Chemicals

Product Code. 11369648
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11369648

Brand: Thermo Scientific Alfa Aesar 044149.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Aluminum oxalate is used as a mordant for printing textiles and dyeing cotton. It finds use as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-87-9
Molecular Formula C6Al2O12
Molecular Weight (g/mol) 318.02
MDL Number MFCD00050791
InChI Key ZCLVNIZJEKLGFA-UHFFFAOYSA-H
Synonym aluminum oxalate, dialuminium trioxalate, unii-dq79q766vn, aluminum, tris ethanedioato 2-di, dialuminum oxalate, aluminum oxalate hydrate, dialuminium 3+ trioxalate ion, dialuminium 3+ trioxalate, aluminum oxalate hydrate 10g, dialuminium 3+ ion trioxalateShow MoreShow Less
PubChem CID 93561
IUPAC Name dialuminum;oxalate
SMILES [Al+3].[Al+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

Color White
Linear Formula Al2(C2O4)3·xH2O
Quantity 250 g
Merck Index 14,355
Solubility Information Insoluble in water.
Formula Weight 318.02 (Anhydrous)
Percent Purity 99%
Physical Form Chunks and/or Powder
Chemical Name or Material Aluminum oxalate hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP14L048 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog Number044149.A1,044149.18,044149.09,044149.30
Lot NumberR25J024 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number044149.A1,044149.18,044149.09,044149.30
Lot NumberU09J036 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number044149.A1,044149.18,044149.09,044149.30
Lot NumberT13J002 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number044149.A1,044149.18,044149.09,044149.30
Lot NumberQ12H040 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044149.A1,044149.18,044149.09,044149.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum oxalate hydrate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.