Learn More
Invitrogen™ RNAlater™-ICE Frozen Tissue Transition Solution
RNAlater-ICE is a novel reagent for transitioning frozen tissue to a state that is easily processed for extraction of high quality RNA.
Brand: Invitrogen™ AM7030
78.67 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
RNAlater-ICE is a novel reagent for transitioning frozen tissue to a state that is easily processed for extraction of high quality RNA. Frozen tissues are simply submerged in RNAlater-ICE and allowed to thaw overnight at -20°C. Once thawed, the tissues can be processed like fresh tissues using standard RNA isolation procedures. No more laborious grinding of frozen tissue to safeguard the RNA.
• Process previously frozen tissues like freshly harvested samples
• Thawing tissues in RNAlater-ICE protects RNA from degradation
• No more tissue pulverizing with mortar and pestle and awkward transfer of powder to tube
• Easily apportion frozen tissue samples for multiple experiments
• Quick Freezing Tissues Preserves RNA
Often, tissues that need to be stored prior to RNA isolation are “snap” or “flash” frozen on dry ice or in liquid nitrogen to preserve RNA integrity. RNA in tissue is stable while frozen at -80°C but thawing the tissue prior to or during its disruption can result in RNA degradation. This is true even if the tissue thaws while in the denaturation solution.
Processing Frozen Tissues is Problematic
Frozen tissues are typically ground with a chilled mortar and pestle in order to maintain RNA quality. Liquid nitrogen must be added to the mortar to keep the sample frozen while it is ground. For multiple samples this process is laborious. Either a separate mortar and pestle set is needed for each sample, or the set must be thawed and cleaned after each sample is processed to avoid cross-contamination. Powdered tissue can also thaw during transfer to a homogenization vessel. This often results in formation of clumps that do not readily disperse in the lysis solution, resulting in RNA degradation and loss. Very small samples should be homogenized immediately in lysis solution, which can again be cumbersome if there are multiple samples to process.
Process Frozen Tissue Without Jeopardizing RNA Integrity
RNAlater-ICE solves all of these problems. Simply submerge frozen tissue samples in 10 volumes of RNAlater-ICE and store overnight at -20°C (the solution will remain liquid at these temperatures). As the tissue thaws, RNA integrity is protected. Once treated, tissue can be safely stored at 4°C or even at room temperature (for a limited period of time) and can be further dissected or processed prior to homogenization in a standard RNA isolation lysis buffer. Thus the same frozen tissue sample can be used multiple times for different experiments without compromising RNA integrity.

Specifications
25 mL RNAlater-ICE Solution; room temperature |
|
Tissue (Frozen) | |
Room Temperature |
RNA Storage Reagent | |
25 mL |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Frozen Tissue Transition Solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H371-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, methanol, isopropanol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
More Discovery, Less Cost
Save on thousands of eligible products.Promo Code: 24111Expires: 2025-03-21
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.