missing translation for 'onlineSavingsMsg'
Learn More

AMPSO, 0.2M buffer soln., pH 8.5, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J61238.AE

44.70 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15463849

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

102029-60-7
0.2 M
Liquid
Odorless
Colorless
8.5
100 mL
249.257
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ25F502 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberZ06D510 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberF27X515 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberZ06D509 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberR16F503 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
AMPSO, 0.2M buffer soln., pH 8.5, Thermo Scientific™ > 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.