missing translation for 'onlineSavingsMsg'
Learn More

Analtech Glass Thin Layer Chromatography (TLC) Developing Tank

Glass Thin Layer Chromatography (TLC) Developing Tank designed for the development of TLC plates.

Brand:  Analtech 34105-50

 View more versions of this product

Product Code. 11770209

  • £247.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: TLC Tank only. (Rack is sold separately; see 34105-25).
This item is not returnable. View return policy

Description

Description

  • Tanks are constructed of durable glass and racks are constructed of anodized aluminum
  • Tanks accommodate standard TLC plates
  • The top surface is ground to a precise tolerance for tight sealing to enable tank saturation and optimal plate development
  • Thinline™ tanks accommodate two plates and require less solvent for development
  • Latch-Lid™ units hold lids in place with stainless steel latches
  • Order rack to develop maximum number of plates in a tank
Note: Requires use of rack. (Order separately; see 34105-25.) If needed, tank with LatchLid™ is Mfr. No. 34105-20.
Specifications

Specifications

Glass
TLC Tank
200 x 200 mm
200 mm
200 mm
1 Ea.
Five 20 x 20 cm Plates
7.9 in.
7.9 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Analtech Glass Thin Layer Chromatography (TLC) Developing Tank > TLC Developing Tank; Size: 200 x 200 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.