missing translation for 'onlineSavingsMsg'
Learn More
Ansell HyFlex™ 11-800 Foam Nitrile Coated Work Gloves Product Code.: 11716723
Estimated Shipment: 20-05-2025
to see stock.
Add to basket

Ansell HyFlex™ 11-800 Foam Nitrile Coated Work Gloves
GREENER_CHOICE

Product Code. 11716723 Shop All Ansell Products
10
Click to view available options
Size:
6
7
9
10
11
This item is not returnable. View return policy

Product Code. 11716723

Brand: Ansell 11800100

Estimated Shipment: 20-05-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Foam nitrile coating enhances grip capabilities which allows workers to easily perform tactile tasks. Gloves are tailored at the base of the pinky finger to better conform to the natural contour of the hand.

  • Ansell ZONZ™ Comfort Fit technology for enhanced fit and improved feel
  • New design provides more durability than previous formulations
  • Foam coating increases breathability for user comfort
  • Proprietary washing process efficiently extracts impurities to deliver a cleaner, more comfortable glove
  • Enhanced grip capabilities in dry and slightly oily environments
Greener Choice Claims
  • Environmental benefits include:
    • Free-of a substance that causes environmental harm
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Palm, Fingers
Coating Material Nitrile Foam
Color Gray, White
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Nylon
Product Type Seamless Knit Gloves
Size 10
For Use With (Application) Automotive Repairs, Heavy Equipment Operation, Metal Fabrication, and Light Chemical Handling
Glove Cuff Style Knit
Packaging Type Polybag
Series 11-800
Show More Show Less
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131

    Expires2025-06-08

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell HyFlex™ 11-800 Foam Nitrile Coated Work Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.