missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq Gold™ 360 Master Mix
GREENER_CHOICE

AmpliTaq Gold 360 PCR Master Mix contains everything required for successful PCR amplification in one convenient package–all components are premixed and premeasured.

Brand:  Applied Biosystems™ 4398881

280.65 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.


 View more versions of this product

Product Code. 10289234

  • £374.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: AmpliTaq Gold 360 Master Mix is supplied at 2X concentration and the 360 GC Enhancer. Typically, 1.5mL of the 360 GC Enhancer is supplied with each 5mL of master Mix.. Source: Recombinant, modified form of theThermus aquaticusDNA polymerase gene expressed in E. coli. All solutions are prepared in deionized, filter-sterilized water.
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

• 2X AmpliTaq Gold 360 Master Mix, 1 x 5 mL
• 360 GC Enhancer, 1 x 1.5 mL

Store at -20°C until first use, then store at 4°C.
High
Standard
Sustainable packaging
5 mL
Cloning, Hot-start PCR
1X
200 Reactions
SuperMix or Master Mix
DNA
Tube
AmpliTaq Gold 360 DNA Polymerase
5' - 3'
PCR Master Mix
Room Temp or Wet Ice
2X
Built-In Hot Start
3'-A
5 kb or less
5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number91326388 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4398881
Lot Number2798669 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4398881
Lot Number2809032 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4398881
Lot Number2798521 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4398881
Lot Number2822990 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4398881
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ AmpliTaq Gold™ 360 Master Mix > 5 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.