Learn More
Applied Biosystems™ MicroAmp™ 12-Cap Strip
Designed to fit on MicroAmp reaction tubes, tube strips, and 96-well plates
Brand:  Applied Biosystems™ N8010534
 View more versions of this product
Product Code. 10498444
  • £82.25 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
MicroAmp™ Cap Strips have a domed profile and are suitable for end-point PCR. 12-cap strips available in clear or an assorted set of red, orange, blue, and green.
  • Will not interfere with sample reading
  • Provides tight seal to minimize evaporation
  • Easy to apply and remove
Minimizes Evaporation
  • Use MicroAmp Caps Strips to seal tubes and reduce possibility of contamination in sample wells and help ensure reliable results
  • Easy to Use
  • Use MicroAmp Cap Installing Tool (sold separately) to push caps down and form tight seal
  • When ready, just peel away caps using tabs at end
  • Colored for Easy Organization
  • Available in different colors to help track tubes during assay preparation
See user's manual or package insert for limited label license and trademark information.
PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Certified DNA/Rnase Free
200 Strips
MicroAmp™ 
Includes 200 MicroAmp™ 12-Cap Strips, Clear. Store at room temperature.
Room Temperature
96
12-Strip Cap
Assorted
Thermal Cyclers
Polypropylene
Standard
Domed Cap
200 Strip Caps
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberI5013Q215 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog Number
Lot NumberI2013Q213 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog Number
Lot NumberI5013Q211 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog Number
Lot NumberI5013Q213 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog Number
Lot NumberI5013Q321 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog Number
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ MicroAmp™ 12-Cap Strip > 200 strips