Learn More
Sartorius™ arium™ Pre-Treatment Cartridge
For the filtration of water in lab water systems.
Brand:  Sartorius™ H2O-CPFAD-1
368.43 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 15648938
  • £403.52 / Each
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory item
  • For use with arium lab water systems
  • Aids in applications such as HPLC, ion chromatography, TOC analysis, PCR, electrophoresis, photometry and many more
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Elix, RiOs
Activated Carbon
Lab water filtration
Cartridge
Polypropylene
Activated Carbon
arium™ systems
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius™ arium™ Pre-Treatment Cartridge > Prefilter cartridge