Learn More
Thermo Scientific™ Ponceau S Staining Solution
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot.
Brand:  Thermo Scientific™ A40000279
45.03 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
 View more versions of this product
Product Code. 17435103
  • £59.25 / 500mL
Explore more special offers
Description
Description
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.
Features include:
• A nondestructive, reversible method to stain and detect proteins on nitrocellulose and PVDF membranes
• Minimal nonspecific interactions with the membrane surface; provides a reliable method for visualization of protein transfer to a membrane
• Simple and quick staining protocol that results in reddish-pink bands that are easily photographed
• Reversed easily in less than five minutes
• Subsequent western blot detection unaffected as the stain does not alter the membrane-bound protein
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Colorimetric
Liquid
Western Blotting
Ponceau S Staining Solution
0.1% (w/v) in 5% acetic acid
500 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZH4428291 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number
Lot NumberZH4368911 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog Number
Lot NumberZD4227421 Certificate TypeCertificate of Analysis Date15/06/2024 Catalog Number
Lot NumberZB4183741 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog Number
Lot NumberSPECS Certificate TypeCertificate of Analysis Date02/03/2024 Catalog Number
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Ponceau S Staining Solution > Ponceau S Staining Solution, 500 mL