missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Coomassie Brilliant Blue Dyes

Thermo Scientific Pierce Coomassie Brilliant Blue dyes are composed of one of the most common forms of coomassie dye, which is a key component of various colorimetric protein gel stains.

Brand:  Thermo Scientific™ 20278

140.60 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 11876744

  • £150.00 / 50g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Pierce Coomassie Brilliant Blue dyes are composed of one of the most common forms of coomassie dye, which is a key component of various colorimetric protein gel stains. Coomassie R-250 and G-250 dyes are two chemical forms of a disulfonated triphenylmethane compound that is commonly used as the basis of stains for detection of proteins in gel electrophoresis and Bradford-type assay reagents for protein quantitation. The R-250 (red-tinted) form lacks two methyl groups that are present in the G-250 (green-tinted) form, which is also called colloidal coomassie dye.

Coomassie R-250 and G-250 dyes are two chemical forms of a disulfonated triphenylmethane compound that is commonly used as the basis of stains for detection of proteins in gel electrophoresis and Bradford-type assay reagents for protein quantitation. The R-250 (red-tinted) form lacks two methyl groups that are present in the G-250 (green-tinted) form, which is also called colloidal coomassie dye.

Typically, coomassie gel stains and protein assay reagents are formulated as very acidic solutions in 25 to 50% methanol. In acidic conditions, the dye binds to proteins primarily through basic amino acids (primarily arginine, lysine, and histidine), and the number of coomassie dye ligands bound to each protein molecule is approximately proportional to the number of positive charges found on the protein. Protein binding causes the dye to change from reddish-brown to bright blue (absorption maximum equals 595 nm).

Features include:
Easy detection—develops intensely colored complexes with proteins
High sensitivity—can determine as little as 0.5 μg/cm2 of protein present in a gel matrix
Reversible staining—anion of Coomassie Brilliant Blue dye formed in the acidic staining medium combines with the protonated amino groups of proteins by electrostatic interaction; resulting complex is reversible under proper conditions
Differentiation between bound and unbound dye—when dissolved in 0.01 M citrate buffer at pH 3.0, has an absorption maximum at 555 nm; protein-dye complex is characterized by a peak slightly broader than that of free dye with a maximum at 549 nm

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature.
Colorimetric
Protein Gel Stain
Pierce
Protein
In-Gel Detection, In-Solution Detection, In-Blot Detection
Coomassie
Brilliant Blue R-250 Dye
50 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZH402435 Certificate TypeCertificate of Analysis Date29/08/2024 Catalog Number20278
Lot NumberZH402435A Certificate TypeCertificate of Analysis Date29/08/2024 Catalog Number20278
Lot NumberZG400549 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number20278
Lot NumberZG400549A Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number20278
Lot NumberZF392859A Certificate TypeCertificate of Analysis Date04/07/2024 Catalog Number20278
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Coomassie Brilliant Blue Dyes > Coomassie Brilliant Blue R-250 Dye

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.