Learn More
Masterflex™ Celcon™ Ratchet Tubing Clamps
Use these durable plastic clamps with flexible tubing. Masterflex™ Celcon™ Ratchet Tubing Clamps won't rust, corrode or distort.
Brand: Masterflex™ 06833-00
This item is not returnable.
View return policy
Description
Twelve-position ratchet control adjusts from full flow to complete closure. Autoclavable.
Specifications
Autoclavable | |
25/Pk. | |
Tubing Clamp | |
For up to 1/2 in. tubing |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Masterflex™ Celcon™ Ratchet Tubing Clamps > For up to 1/2 in. tubing.
Spot an opportunity for improvement?Share a Content Correction