Learn More
Azlon™ Polyethylene Stoppers
Use these stoppers with glass or plastic volumetric flasks and measuring cylinders which have longer 'B' length sockets. Azlon™ Polyethylene Stoppers feature a large head for a good grip and extra protection if glassware is knocked over.
Brand: Azlon™ TRF592
17.96 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Use these stoppers with glass or plastic volumetric flasks and measuring cylinders which have longer 'B' length sockets. AzlonTM Polyethylene Stoppers feature a large head for a good grip and extra protection if glassware is knocked over.
- Manufactured from polyethylene
- Available in white and red
- Suitable for use with both Azlon and glass volumetric ware
- Enlarged head provides a good grip and protects glassware
- Ribbed cone ensures a secure leak free seal
Specifications
Glass or Plastic volumetric flasks and measuring cylinders etc | |
White, Red | |
Tapered Stopper |
12/21 | |
Polyethylene | |
Ribbed Cone |
Product Suggestions
Customers who viewed this item also viewed