Learn More
Saint Gobain Life Sciences Versilic™ Folding Skirt Stoppers
Silicone biologically neutral Versilic™ Folding Skirt Stoppers.
Brand: Saint Gobain Life Sciences 467030-05
Description
- Provides complete tightness on glass, metal or plastic containers regardless of their rims.
- The turnover is molded to grip the outside of the container neck, providing a double seal in conjunction with the annular serrations on the hollow plug.
- Can be perforated.
- Remains watertight after several perforations.
- Recommended for biological applications.
- Temperatures between -60°C and +220°C.
Specifications
27.6 to 36.3 mm | |
-60°C to +200°C | |
Silicone Stopper |
5 Pk. | |
Silicone |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Saint Gobain Life Sciences Versilic™ Folding Skirt Stoppers > Versilic™ Folding Skirt Stoppers; Inner Diameter: 27.6 to 36.3 mm
Spot an opportunity for improvement?Share a Content Correction