missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ B-PER™ Reagent (in Phosphate Buffer)

Efficiently lyse E.coli and other bacterial cells and gently extract native proteins with these nonionic detergent solutions.

Brand:  Thermo Scientific™ 78266

277.34 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


385.70 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11894091

  • £416.00 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

B-PER Bacterial Protein Extraction Reagent (in Phosphate Buffer)
Liquid
500 mL
B-PER™
500 mL
Reagent
Store at room temperature.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZJ386761 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog Number78266
Lot NumberSPECS Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number78266
Lot NumberYF373379 Certificate TypeCertificate of Analysis Date06/07/2023 Catalog Number78266
Lot NumberXD340876 Certificate TypeCertificate of Analysis Date17/05/2022 Catalog Number78266
Lot NumberKA125188 Certificate TypeCertificate of Analysis Date22/03/2022 Catalog Number78266
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ B-PER™ Reagent (in Phosphate Buffer) > Reagent in phosphate buffer; 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.