missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Adjuvants
Used to prepare antigen-adjuvant emulsions. BD™ Difco™ Adjuvants are used in immunological studies with laboratory animals. (Not intended for human or therapeutic use.)
£74.60 - £1255.00
Specifications
Content And Storage | Store in cool, dry conditions in well sealed containers, away from oxidizing agents. |
---|---|
For Use With (Application) | Immunological studies with laboratory animals |
Alert:
Not intended for human or therapeutic use.
Product Code | Brand | Quantity | Product Type | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Product Type | Description | Price | Quantity & Availability | |||
11719062
|
BD
263810 |
6 x 10 mL | Freund's Complete Adjuvant | Suspension of 5mg M. butyricum in a mixture of paraffin oil and an emulsifying agent |
£231.00
60mL |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11729062
|
BD
263910 |
6 x 10 mL | Freund's Incomplete Adjuvant | Adjuvant, Incomplete (Freund) |
£74.60
60mL |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17224363
|
BD
264010 |
6 x 100 mg | Mycobacterium butyricum | Killed M. butyricum for use in adjuvants |
£1255.00
Pack of 6 |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11762934
|
BD
231131 |
6 x 10 mL | Complete H37Ra | Suspension of 10mg M. tuberculosis in a mixture of paraffin oil and an emulsifying agent |
£322.00
Pack of 6 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10218823
|
BD
231141 |
6 x 100 mg | Mycobacterium tuberculosis H37Ra | Killed M. tuberculosis H37 Ra for use in adjuvants |
£1010.00
600mg |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Adjuvants are mixed with aqueous antigen solution or suspension
- Once injected, they provide a pool of emulsified antigen at the injection site and allow the antigen to be released over an extended period of time
- With adjuvants, the immune response to antigens (especially soluble antigens) is enhanced and prolonged
- Use Incomplete Freund adjuvant to avoid the use of acid-fast bacilli; however, the antibody response is not as marked as with the complete adjuvant
Specifications
Store in cool, dry conditions in well sealed containers, away from oxidizing agents. | |
Immunological studies with laboratory animals |