Learn More
Bel-Art™ SP Scienceware™ Equilibrium Dialysis Cells
Feature stainless steel screws that tighten down on the dialysis membrane also acting as a gasket
Brand:  Bel-Art™ H40260-0000
 View more versions of this product
Product Code. 15204091
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Made from clear acrylic
  • Filler hole screws on each half cavity are fluoropolymer-coated stainless steel
  • Volume shown is for one half cell (double for total cell volume)
  • A red dot on one half of the equilibrium cell easily identifies the otherwise identical halves
Specifications
Specifications
Dialysis Cell
1 mL
Each
Acrylic
Transparent
Stainless steel screws that tighten down on the dialysis membrane also acting as a gasket
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bel-Art™ SP Scienceware™ Equilibrium Dialysis Cells > 1mL