missing translation for 'onlineSavingsMsg'
Learn More
Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution, Thermo Scientific Chemicals Product Code.: 10734034

Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution, Thermo Scientific Chemicals

Product Code. 10734034
2.5 L, Glass bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10734034

Brand: Thermo Scientific Acros 263820025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-54-5, 7732-18-5
Molecular Formula C9H13ClN
Molecular Weight (g/mol) 170.66
MDL Number MFCD00145757
Synonym Alkyl-benzyl-dimethylammonium chloride

Specifications

Color Colorless to Yellow
Boiling Point >100.0°C
CAS Min % 49.0, 48.0
CAS Max % 52.0, 51.0
Linear Formula C6H5CH2N(CH3)2RClR=C8H17toC18H37
Concentration 49 to 52% (based on M.W.= 352.5)
Density 0.9800g/mL
Merck Index 15, 1061
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone, slightly soluble in benzene, almost insoluble in ether
Packaging Glass bottle
Specific Gravity 0.98
Viscosity 130 mPa.s (20°C)
pH 6 to 9 (10% aq. soln.)
Quantity 2.5 L
Flash Point 105°C
Physical Form Solution
Chemical Name or Material Benzalkonium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467567 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number263820010,263820025
Lot NumberA0466199 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog Number263820010,263820025
Lot NumberA0465415 Certificate TypeCertificate of Analysis Date14/08/2024 Catalog Number263820010,263820025
Lot NumberA0464457 Certificate TypeCertificate of Analysis Date10/07/2024 Catalog Number263820010,263820025
Lot NumberA0463999 Certificate TypeCertificate of Analysis Date24/06/2024 Catalog Number263820010,263820025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzalkonium chloride, 50 wt% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.