Learn More
MedChemExpress Biotin LC hydrazide
MedChemExpress Biotin LC hydrazide is a long chain protein modification reagent, which can transform periodate-oxidized glycoproteins.
Brand: MedChemExpress HY-101885
This item is not returnable.
View return policy
Specifications
5 mg | |
4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) | |
109276-34-8 | |
DMSO : 31 mg/mL (83.45 mM; Need ultrasonic) |
Biochemical Assay Reagents | |
Room temperature | |
371.5 g/mol | |
Biotinamidocaproyl hydrazide ∣ BACH ∣ (+)-Biotinamidohexanoic Acid hydrazide ∣ Biotin Aminocaproyl Hydrazide |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
MedChemExpress Biotin LC hydrazide
Spot an opportunity for improvement?Share a Content Correction