missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Blotting Paper, Grade BF 4, Sheets

Sartorius Stedim Biotech™ Blotting Paper, Grade BF 4, Sheets are highly pure and ideal for blotting chromatography.

Brand:  Sartorius Stedim Biotech™ FT-2-521-150150G

44.40 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15912810

  • £46.20 / Pack of 25
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • This highly pure paper grade is not only ideal for blotting and chromatography, but also for a wide range of absorption applications like those common in the life sciences and diagnostics.
  • Made of high-purity cotton linters for uniform buffer flow and resulting blots.
  • No additives to avoid any interference during transfer.
  • BF 4 grade is ideal for transferring DNA or RNA according to the Southern technique or semidry blotting of proteins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Capillary rise: 160mm/10 min., Weight: 550 g/m2, 100/Pk
1.30 mm
BF 4
150 mm
150 x 150 mm
Sheet
150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 4, Sheets > 150 x 150mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.