missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bovine Pituitary Extract (BPE)

Bovine Pituitary Extract (BPE)

Brand:  Gibco™ 13028014

Product Code. 11568866

  • £66.50 / 25mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

The protein concentration of each lot of Bovine Pituitary Extract is determined by the Bradford method. Each vial of BPE contains not less than 25 mg of protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5 to -30°C).
Mammalian Cell Culture
Bovine Pituitary Extract
25 mg
Mammalian Cells
Liquid
6 to 8
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2821029 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number13028014
Lot Number2864953 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number13028014
Lot Number2864954 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number13028014
Lot Number2864955 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number13028014
Lot Number2981708 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number13028014
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Bovine Pituitary Extract (BPE) > 25mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.