Learn More
Leica Microsystems DM500 Brightfield Microscope
Ideal tool to make teaching entry-level college life science courses easy and fun for instructor and student. DM500 Brightfield Microscope has plug-and-play capability.
Brand: Leica Microsystems 1050DM5002
This item is not returnable.
View return policy
Description
All systems include XY mechanical stage, coarse/fine focus, dust cover, immersion oil and user manual.The ideal tool for making life science university courses easy and fun for both instructor and student
- The pre-focused, pre-centred condenser and the EZTube™ preset dioptre eyepieces prevent incorrect adjustments and provide more time for hands-on teaching
- EZStore™ design with integrated handle and cord wrap allows easy carrying and lifting, and protection against microscope component damage
- Features LED illumination, eliminating the nuisance and expense of replacing bulbs
- The optical system uses the same platform as Leica research grade microscopes
- 'Rackless' stage system means there is no rack and pinion mechanism protruding from the side of the stage - this prevents minor scratches or grazes to the skin, or the tearing of gloves
Specifications
Prefocused, precentered Abbe condenser | |
LED | |
DM5002 | |
Brightfield Microscope | |
220/240 V 50/60 Hz |
Binocular, High Eyepoint | |
4X, 10X, 40X, 100X (oil) and 10X/18 pointer | |
Brightfield | |
219 x 282 x 428 mm (standard configuration) |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Product Title
Leica Microsystems DM500 Brightfield Microscope >
Spot an opportunity for improvement?Share a Content Correction