missing translation for 'onlineSavingsMsg'
Learn More
Brucine, Thermo Scientific Chemicals Product Code.: 15483799

Brucine, Thermo Scientific Chemicals

Product Code. 15483799
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15483799

Brand: Thermo Scientific Alfa Aesar J61178.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Brucine is used as a denaturing alcohol, lubricant additive, separation of racemic mixtures. Proposed to reduce inflammatory mediators including COX-2, PGE2 and thromboxane B2. Able to induce cell apoptosis, with cytotoxic and antiproliferative effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brucine is used as a denaturing alcohol, lubricant additive, separation of racemic mixtures. Proposed to reduce inflammatory mediators including COX-2, PGE2 and thromboxane B2. Able to induce cell apoptosis, with cytotoxic and antiproliferative effects.

Solubility
Soluble in alcohol, chloroform, and benzene, slightly soluble in water, ether, and glycerol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 357-57-3
Molecular Formula C23H26N2O4
Molecular Weight (g/mol) 394.471
MDL Number MFCD00005942
InChI Key RRKTZKIUPZVBMF-VLTSECPOSA-N
Synonym brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy
PubChem CID 51413923
IUPAC Name (4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-oneShow MoreShow Less
SMILES COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC

Specifications

Melting Point 175°C to 178°C (decomposition)
Color White
Quantity 100 g
UN Number UN1570
Beilstein 63046
Merck Index 14,1455
Solubility Information Soluble in alcohol,chloroform,and benzene,slightly soluble in water,ether,and glycerol
Optical Rotation −118° (c=2.5 in Chloroform)
Formula Weight 394.47
Physical Form Powder
Chemical Name or Material Brucine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS06J002 Certificate TypeCertificate of Analysis Date13/06/2023 Catalog NumberJ61178.22
Lot NumberS14I037 Certificate TypeCertificate of Analysis Date21/06/2022 Catalog NumberJ61178.06,J61178.14,J61178.22
Lot NumberW13H025 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberJ61178.06,J61178.14,J61178.22
Lot NumberP18H035 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberJ61178.06,J61178.14,J61178.22
Lot NumberL16Z001 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ61178.06,J61178.14,J61178.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brucine, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.