missing translation for 'onlineSavingsMsg'
Learn More
Brucine, 99%, anhydrous, Thermo Scientific Chemicals Product Code.: 10675511

Brucine, 99%, anhydrous, Thermo Scientific Chemicals

Product Code. 10675511
100 g, Glass bottle
Click to view available options
Quantity:
5 g
50 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10675511

Brand: Thermo Scientific Acros 278721000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 357-57-3
Molecular Formula C23H26N2O4
Molecular Weight (g/mol) 394.46
MDL Number MFCD00005942
InChI Key RRKTZKIUPZVBMF-VLTSECPOSA-N
Synonym brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy
PubChem CID 51413923
IUPAC Name (4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-oneShow MoreShow Less
SMILES COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC

Specifications

Melting Point 174.0°C to 179.0°C
Color White to Beige
Loss on Drying 1% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 100 g
Beilstein 27, IV, 7876
Merck Index 15, 1464
Specific Rotation − 123.00
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, chloroform and benzene, slightly soluble in ether, glycerol and, ethyl acetate
Formula Weight 394.46
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Brucine, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0433032 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog Number278721000,278720050,278725000,278720500
Lot NumberA0453194 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number278721000,278720050,278725000,278720500
Lot NumberA0397660 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number278721000,278720050,278725000,278720500
Lot NumberA0403511 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number278721000,278720050,278725000,278720500
Lot NumberA0412115 Certificate TypeCertificate of Analysis Date22/03/2022 Catalog Number278721000,278720050,278725000,278720500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brucine, 99%, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.