Learn More
BTX™ HT 25-well Electroporation Plates
HT 25-well Electroporation Plates for use with ECM 630 and ECM 830 Electroporation Systems.
Brand: BTX™ 450462
Description
- 25-well disposable electroporation plate
- Standard multi-well formats
- Bio-compatible materials
Specifications
25-well | |
ECM 830 HT Electroporation System, the ECM 630 HT Electroporation System, or the Gemini X2 HT Electroporation System | |
BTX Electroporation | |
25-Well Disposable Electroporation Plate, 4 mm gap, 200 μl, 1 plate | |
1 Ea. | |
4 mm | |
Sterile | |
Yes |
Transfection/transformation of bacteria, plant and mammalian cells, siRNA Transfection, Library Studies and Stem Cell Project | |
Medical grade polycarbonate, polished aluminum | |
Electroporation consumables | |
25-well Electroporation Plate | |
0.15 in. | |
200 μL | |
Exponential Decay Wave, Square Wave |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.