Description
- Soft, light non-woven polypropylene with an elasticated edge
- Complete hair coverage
- Ultra low-linting, making them ideally suited to working environments where contamination by loose fibers may be a risk
- Food safe and Latex-free for reduced irritation
- Larger than a mob cap and designed for longer hair
- One size fits all
Specifications
Specifications
Product Type | Bouffant |
Size | One Size |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Latex Free | Yes |
Model | D84110 |
Quantity | 1000/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Product Title
Pal International™ Bouffant Caps >
Spot an opportunity for improvement?Share a Content Correction