missing translation for 'onlineSavingsMsg'
Learn More
Calcium phosphate, dibasic, 98+%, for analysis, anhydrous, Thermo Scientific Chemicals Product Code.: 12675027

Calcium phosphate, dibasic, 98+%, for analysis, anhydrous, Thermo Scientific Chemicals

Product Code. 12675027
50 g, Plastic bottle
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 12675027

Brand: Thermo Scientific Acros 446430500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-93-9
Molecular Formula CaHO4P
Molecular Weight (g/mol) 136.06
MDL Number MFCD00010909
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
Synonym calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1Show MoreShow Less
PubChem CID 24441
ChEBI CHEBI:32596
IUPAC Name calcium;hydrogen phosphate
SMILES [Ca++].OP([O-])([O-])=O

Specifications

Quantity 50 g
Assay Percent Range 98.0 to 103% (Complexometry)
Linear Formula CaHPO4
Merck Index 15,1694
Solubility Information Solubility in water: 0.1g/L (25°C). Other solubilities: practically insoluble in alcohol
Formula Weight 136.06
Percent Purity ≥98%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Calcium phosphate, dibasic

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466826 Certificate TypeCertificate of Analysis Date23/10/2024 Catalog Number446432500,446430500
Lot NumberA0465417 Certificate TypeCertificate of Analysis Date26/08/2024 Catalog Number446432500,446430500
Lot NumberA0462728 Certificate TypeCertificate of Analysis Date22/05/2024 Catalog Number446432500,446430500
Lot NumberA0460689 Certificate TypeCertificate of Analysis Date14/03/2024 Catalog Number446432500,446430500
Lot NumberA0460200 Certificate TypeCertificate of Analysis Date15/02/2024 Catalog Number446432500,446430500
Product Identifier
  • Calcium phosphate, dibasic

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium phosphate, dibasic, 98+%, for analysis, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.