missing translation for 'onlineSavingsMsg'
Learn More
Capsazepine, 98+%, Thermo Scientific™ Product Code.: 15405329

Capsazepine, 98+%, Thermo Scientific™

Product Code. 15405329
10mg
Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy

Product Code. 15405329

Brand: Thermo Scientific Alfa Aesar J63055.MA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A competitive antagonist for capsaicin and resiniferatoxin

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Capsazepine is shown to inhibit the development of mechanical hyperalgesia induced by intraplanar capsaicin injection. Capsazepine is described to block Resiniferatoxin (sc-24015)- and Capsaicin-induced contractions of guinea pig treacheal smooth muscle. Studies investigating the function of the VR1 (TRPV1 receptor) have employed Capsazepine as a tool for perturbing systemic neuronal response to Capsaicin.

Solubility
Soluble in ethanol or DMSO

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138977-28-3
Molecular Formula C19H21ClN2O2S
Molecular Weight (g/mol) 376.899
MDL Number MFCD00153778
InChI Key DRCMAZOSEIMCHM-UHFFFAOYSA-N
Synonym capsazepine, capsazepin, n-2-4-chlorophenyl ethyl-1,3,4,5-tetrahydro-7,8-dihydroxy-2h-2-benzazepine-2-carbothioamide, unii-lfw48my844, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide, 2-2-4-chlorophenyl ethylamino-thiocarbonyl-7,8-dihydroxy-2,3,4,5-tetrahydro-1h-2-benzazepine, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2h-2-benzazepine-2-carbothioamide, capasazepine, tocris-0464
PubChem CID 2733484
ChEBI CHEBI:70773
IUPAC Name N-[2-(4-chlorophenyl)ethyl]-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide
SMILES C1CC2=CC(=C(C=C2CN(C1)C(=S)NCCC3=CC=C(C=C3)Cl)O)O

Specifications

Quantity 10mg
Solubility Information Soluble in ethanol or DMSO
Formula Weight 376.9
Percent Purity ≥99%
Chemical Name or Material Capsazepine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ21E031 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ63055.MA,J63055.MF
Lot NumberP24B042 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ63055.MA,J63055.MF
Lot NumberM29F043 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ63055.MA,J63055.MF
Lot NumberY05F098 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ63055.MA,J63055.MF
Lot NumberX31F043 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ63055.MA,J63055.MF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Capsazepine, 98+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.