missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium fluoride, 99.9%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent

Specifications
Specifications
Boiling Point | 1251.0°C |
Melting Point | 682.0°C |
Color | White |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.9% |
Linear Formula | CsF |
Fieser | 01,121; 06,100; 07,57; 08,81; 09,100; 10,81; 11,115; 12,108; 13,68; 14,79; 15,75; 16,69; 17,68 |
Merck Index | 15, 2011 |
Solubility Information | Solubility in water: 369g/100mL (18°C). Other solubilities: soluble in methanol, insoluble in dioxane and pyridine |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0443011 | Certificate of Analysis | 31/01/2025 | 315910050,315911000,315910250 |
A0461137 | Certificate of Analysis | 18/03/2024 | 315910050,315911000,315910250 |
A0395001 | Certificate of Analysis | 14/12/2022 | 315910050,315911000,315910250 |
A0443011 | Certificate of Analysis | 10/06/2022 | 315910050,315911000,315910250 |
A0431073 | Certificate of Analysis | 08/06/2021 | 315910050,315911000,315910250 |
Safety and Handling




Product Identifier
- Cesium fluoride
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H361-Suspected of damaging fertility or the unborn child.
- EUH032-Contact with acids liberates very toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cesium fluoride, 99.9%, extra pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction