Learn More
Thermo Scientific™ Chromacol™ GOLD-Grade Inert Vials and Inserts
Ensure sample security and integrity with inert vials that have the highest recovery rates, an ultra-low pH shift, and lowest alkali content.
Brand: Thermo Scientific™ 03-CVG
Description
Maximize your sample performance and have fewer failed assays with Thermo Scientific™ Chromacol™ GOLD-Grade Inert Glass Vials. These vials offer the most state-of-the-art technology in the industry for the analysis of critical low detection limit analytes, and when sample quality, reproducibility, security, and integrity are of upmost importance.
For easier sample handling and less risk of sample loss, the Chromacol GOLD-Grade Vials are the best-in-class choice for all chromatography users performing ultra-trace analysis.
Choose the right vial for your needs- 8mm crimp top seal minimizes exposure between sample solvent and septum
- Neutral Borosilicate GOLD-Grade glass is a low expansion, high purity glass with an extremely low concentration of active sites to protect your analytes from unwanted interactions
- Precision-machined (glass or PTFE) sleeves allow the vials to be used in the vast majority of commercial autosamplers
Thermo Scientific™
Agilent™
Beckman™
Carlo Erba™
CTC™
Fisons™
PerkinElmer™
Shimadzu™
VWR™ (Merck)/Hitachi™
*See the Thermo Scientific Columns and Consumables Catalog for autosampler compatibility

Specifications
0.3mL Crimp Top, Round Bottom Vial, Clear Gold Grade | |
6 mm | |
325 μL | |
Chromacol Gold | |
500 | |
8 mm |
Clear | |
Glass | |
<5 mL | |
32 mm | |
Crimp Top Vial | |
No |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.