missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Chrysoidin, pure Product Code.: 10153880

Thermo Scientific Chemicals Chrysoidin, pure

Product Code. 10153880
50 g, Glass bottle
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10153880

Brand: Thermo Scientific Chemicals 200740500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 532-82-1 | C12H12N4·HCl | 248.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 532-82-1
Melting Point 235.0°C
Color Bordeaux to Purple
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C12H12N4·HCl
MDL Number MFCD00012976
Quantity 50 g
Beilstein 16, 383
Merck Index 15, 2261
Synonym chrysoidine, basic orange 2, chrysoidine g, chrysoidine y, chrysoidin, c.i. basic orange 2, chrysoidine ygh, chrysoidine a, chrysoidine b, chrysoidine j
Solubility Information Solubility in water: 55g/L (15°C). Other solubilities: 4.75% absolute ethanol - 6.0% cellosolve, 9.5% anhydr.ethylene glycol, slightly soluble in acetone, practically insoluble in benzene, xylene and ether
InChI Key MCTQNEBFZMBRSQ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)N=NC2=C(C=C(C=C2)N)N.Cl
IUPAC Name 4-phenyldiazenylbenzene-1,3-diamine;hydrochloride
Molecular Weight (g/mol) 248.72
PubChem CID 10771
Formula Weight 248.72
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material Chrysoidin
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0441563 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog Number200740500
Lot NumberA0374196 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number200740500
Lot NumberA0401530 Certificate TypeCertificate of Analysis Date28/04/2021 Catalog Number200740500
Lot NumberA0420263 Certificate TypeCertificate of Analysis Date08/07/2020 Catalog Number200740500
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chrysoidin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Chrysoidin, pure >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.