Learn More
Hirschmann™ Class B Measuring Cylinders, Blue Stain Graduation
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class B Measuring Cylinders, Blue Stain Graduation are manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 2230190
 View more versions of this product
Product Code. 11788021
  • £99.35 / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With spout, short line
  • Blue stain graduation
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Hexagonal
Round
B
±3.75 mL
Glass
5 mL
Blue
DIN EN ISO 4788
390 mm
Graduated Measuring Cylinder
500 mL
3.75
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hirschmann™ Class B Measuring Cylinders, Blue Stain Graduation > Capacity: 500mL; Height: 390mm; Blue Graduations; Hexagonal Glass Socket