missing translation for 'onlineSavingsMsg'
Learn More
Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10414711

Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10414711
100 g, Plastic Bottle
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10414711

Brand: Thermo Scientific Acros 423571000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Melting Point 86°C
Color Lilac
Physical Form Fine Crystalline Powder
Quantity 100 g
Assay Percent Range 98.0 to 102% (Complexometry)
Linear Formula CoCl2·6H2O
Fieser 01,155; 06,127; 11,479
Merck Index 15, 2421
Solubility Information Solubility in water: 970g/L (20°C)
Formula Weight 237.93
Percent Purity 98 to 102%
Grade ACS Reagent
Packaging Plastic Bottle
Vapor Pressure Negligible
Chemical Name or Material Cobalt(II) chloride hexahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468018 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number423571000,423575000,423570050
Lot NumberA0445338 Certificate TypeCertificate of Analysis Date23/02/2024 Catalog Number423571000,423575000,423570050
Lot NumberB0145388R Certificate TypeCertificate of Analysis Date20/10/2023 Catalog Number423571000,423575000,423570050
Lot NumberB0147436 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog Number423571000,423575000,423570050
Lot NumberB0155240 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog Number423571000,423575000,423570050
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H341-Suspected of causing genetic defects.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.