Learn More
R&D Systems™ CometAssay Electrophoresis System
Electrophoresis System for the CometAssay™
Brand:  R&D Systems™ 4250-050-ES
Product Code. 30001322
  • £2600.00 / 1 set
Explore more special offers
Alert:
Caution! Electrical Hazard! This equipment is designed for use with a DC power supply providing up to 250 VDC. Careless handling can result in electrical shock. To avoid any risk of injury, the instrument should be operated in accordance with the instruction provided.
Description
Description
  • Overcomes assay variability first by placing an acrylic overlay on top of an elevated slide tray to maintain optimal buffer height for DNA migration
  • Secondly, a constant buffer temperature is maintained using an external chamber under the tank to cool the glass slide platform and buffer chamber
  • Electrophoresis time for optimal DNA migration is achieved in 30 minutes with Alkaline (pH>13) Electrophoresis Solution, or 45 minutes with Neutral Electrophoresis Buffer at 1V/cm
  • The smoke grey colored unit is designed to minimize exposure to UV light but still allow visual inspection
  • Third, specially designed slide trays are also provided to accommodate 2, 20 and 96 well slides and maintain proper slide orientation during electrophoresis
  • CometAssay™ Electrophoresis System II, CometAssay™ Alkaline Control Cells, and CometAssay™ Neutral Control Cells enable investigators to consistently optimize their comet assays for highly reproducible results, and to standardize electrophoresis methods between individual users and laboratories
TRUSTED_SUSTAINABILITY
Specifications
Specifications
The product requires storage at both 2 - 8°C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
A simple and effective method for evaluating DNA damage in cells
Electrophoresis Tank with Leveling Bubble, Safety Lid & Cables, 2/20 Well Slide Trays, and Slide Tray Overlay
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ CometAssay Electrophoresis System > Sufficient For: A simple and effective method for evaluating DNA damage in cells