missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ Conductivity Cell with 1m Fixed Cable

SI Analytics™ Conductivity Cell with 1m Fixed Cable is constructed of a platinum sensor and glass shaft.

Brand:  SI Analytics™ 285106189

Product Code. 10661182

  • £620.00 / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 4 pole cell
  • Glass shaft
  • 1m cable with 8 pole plug
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Conductivity Measuring Cell
120 mm
1 m with 8 Pole Plug
12 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SI Analytics™ Conductivity Cell with 1m Fixed Cable > LengthMetric: 120mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.